3-(3,4-Difluorophenyl)propionic acid, 98%
3-(3,4-Difluorophenyl)propionic acid, 98%
3-(3,4-Difluorophenyl)propionic acid, 98%
Thermo Scientific Chemicals

3-(3,4-Difluorophenyl)propionic acid, 98%

CAS: 161712-75-0 | C9H8F2O2 | 186.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26185.03
also known as H26185-03
Price (EUR)
17,90
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
17,90
Each
Chemical Identifiers
CAS161712-75-0
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Melting Point (clear melt)48.0-54.0?C
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only