3-Bromo-5-(trifluoromethyl)benzaldehyde, 97%
3-Bromo-5-(trifluoromethyl)benzaldehyde, 97%
3-Bromo-5-(trifluoromethyl)benzaldehyde, 97%
3-Bromo-5-(trifluoromethyl)benzaldehyde, 97%
Thermo Scientific Chemicals

3-Bromo-5-(trifluoromethyl)benzaldehyde, 97%

CAS: 477535-41-4 | C8H4BrF3O | 253.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26197.03
also known as H26197-03
Price (EUR)
29,30
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
29,30
Each
Add to cart
Chemical Identifiers
CAS477535-41-4
IUPAC Name3-bromo-5-(trifluoromethyl)benzaldehyde
Molecular FormulaC8H4BrF3O
InChI KeyPCRLZGCXLNNMFL-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(Br)=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1.5%
FormLiquid
Refractive Index1.5085-1.5135 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only