4-Chloro-3-fluorobenzyl bromide, 97%
4-Chloro-3-fluorobenzyl bromide, 97%
4-Chloro-3-fluorobenzyl bromide, 97%
4-Chloro-3-fluorobenzyl bromide, 97%
Thermo Scientific Chemicals

4-Chloro-3-fluorobenzyl bromide, 97%

CAS: 206362-80-3 | C7H5BrClF | 223.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26391.03
also known as H26391-03
Price (EUR)
96,40
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
96,40
Each
Add to cart
Chemical Identifiers
CAS206362-80-3
IUPAC Name4-(bromomethyl)-1-chloro-2-fluorobenzene
Molecular FormulaC7H5BrClF
InChI KeyCNUYBEIHDWMLSD-UHFFFAOYSA-N
SMILESFC1=C(Cl)C=CC(CBr)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
Melting Point (clear melt)27.0-37.0?C
Assay (GC)≥96.0%
FormFused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only