3,5-Difluoro-4-methoxyaniline, 97%
3,5-Difluoro-4-methoxyaniline, 97%
3,5-Difluoro-4-methoxyaniline, 97%
Thermo Scientific Chemicals

3,5-Difluoro-4-methoxyaniline, 97%

CAS: 363-47-3 | C7H7F2NO | 159.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26778.06
also known as H26778-06
Price (EUR)
557,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
557,00
Each
Chemical Identifiers
CAS363-47-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)73.5-82.5?C
Assay (GC)≥96.0%
3,5-Difluoro-4-methoxyaniline is used in conjunction with thermal energy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-4-methoxyaniline is used in conjunction with thermal energy.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Carl Niemann.; Andrew A. Benson.; James F. Mead. The Synthesis of 3',5'-Difluoro-dl-thyronine and 3,5-Diiodo-3',5'-difluoro-dl-thyronine. J. Am. Chem. Soc. 1941, 63, (8), 2204-2208.
  2. Jian Qiu.; Scott H. Stevenson.; Michael J. O'Beirn e.; Richard B. Silverman. 2,6-Difluorophenol as a Bioisostere of a Carboxylic Acid: Bioisosteric Analogues of γ-Aminobutyric Acid. J. Med. Chem. 1999, 42, (2), 329-332.