Homopiperazin-5-one, 95%
Homopiperazin-5-one, 95%
Homopiperazin-5-one, 95%
Thermo Scientific Chemicals

Homopiperazin-5-one, 95%

CAS: 34376-54-0 | C5H10N2O | 114.148 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H26990.03
also known as H26990-03
Price (EUR)
249,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
249,00
Each
Specifications
CAS34376-54-0
Chemical Name or MaterialHomopiperazin-5-one
Formula Weight114.14
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only