4-cis-Fluoro-L-prolinamide hydrochloride, 98%
4-cis-Fluoro-L-prolinamide hydrochloride, 98%
4-cis-Fluoro-L-prolinamide hydrochloride, 98%
Thermo Scientific Chemicals

4-cis-Fluoro-L-prolinamide hydrochloride, 98%

CAS: 426844-23-7 | C5H10ClFN2O | 168.596 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27235.06
also known as H27235-06
Price (EUR)
377,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
377,00
Each
Chemical Identifiers
CAS426844-23-7
IUPAC Name4-fluoropyrrolidine-2-carboxamide hydrochloride
Molecular FormulaC5H10ClFN2O
InChI KeyYIEACUBSPQGCLW-UHFFFAOYNA-N
SMILESCl.NC(=O)C1CC(F)CN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Optical Rotation-48.5? ± 0.5? (c=0.5 in methanol)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only