6-Bromo-2H-1,4-benzoxazin-3(4H)-one, 95%
6-Bromo-2H-1,4-benzoxazin-3(4H)-one, 95%
6-Bromo-2H-1,4-benzoxazin-3(4H)-one, 95%
Thermo Scientific Chemicals

6-Bromo-2H-1,4-benzoxazin-3(4H)-one, 95%

CAS: 24036-52-0 | C8H6BrNO2 | 228.045 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H27493.03
also known as H27493-03
Price (EUR)
48,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
48,40
Each
Chemical Identifiers
CAS24036-52-0
IUPAC Name6-bromo-3,4-dihydro-2H-1,4-benzoxazin-3-one
Molecular FormulaC8H6BrNO2
InChI KeyUQCFMEFQBSYDHY-UHFFFAOYSA-N
SMILESBrC1=CC2=C(OCC(=O)N2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (HPLC)≥94.0%
Melting Point (clear melt)219.0-228.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only