2H-1,4-Benzoxazin-3(4H)-one, 99%
2H-1,4-Benzoxazin-3(4H)-one, 99%
2H-1,4-Benzoxazin-3(4H)-one, 99%
Thermo Scientific Chemicals

2H-1,4-Benzoxazin-3(4H)-one, 99%

CAS: 5466-88-6 | C8H7NO2 | 149.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27622.06
also known as H27622-06
Price (EUR)
24,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
24,20
Each
Chemical Identifiers
CAS5466-88-6
IUPAC Name3,4-dihydro-2H-1,4-benzoxazin-3-one
Molecular FormulaC8H7NO2
InChI KeyQRCGFTXRXYMJOS-UHFFFAOYSA-N
SMILESO=C1COC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)170.0-176.0?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only