4,4-Difluoro-L-prolinamide hydrochloride, 98%, Thermo Scientific Chemicals
4,4-Difluoro-L-prolinamide hydrochloride, 98%, Thermo Scientific Chemicals
4,4-Difluoro-L-prolinamide hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4-Difluoro-L-prolinamide hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H27770.03
also known as H27770-03
1 g
Catalog number H27770.03
also known as H27770-03
Price (EUR)
162,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
162,00
Each
Chemical Identifiers
CAS426844-51-1
IUPAC Name4,4-difluoropyrrolidine-2-carboxamide hydrochloride
Molecular FormulaC5H9ClF2N2O
InChI KeyRRQDQYAEZGKHOB-UHFFFAOYNA-N
SMILESCl.NC(=O)C1CC(F)(F)CN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)White to cream to pale brown
Optical Rotation-30.0 ± 2.0? (c=1 in water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only