(+/-)-1-Boc-3-hydroxypyrrolidine, 97%
(+/-)-1-Boc-3-hydroxypyrrolidine, 97%
(+/-)-1-Boc-3-hydroxypyrrolidine, 97%
Thermo Scientific Chemicals

(+/-)-1-Boc-3-hydroxypyrrolidine, 97%

CAS: 103057-44-9 | C9H17NO3 | 187.239 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H28750.06
also known as H28750-06
Price (EUR)
97,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
97,20
Each
Chemical Identifiers
CAS103057-44-9
IUPAC Nametert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
Molecular FormulaC9H17NO3
InChI KeyAPCBTRDHCDOPNY-ZETCQYMHSA-N
SMILESCC(C)(C)OC(=O)N1CC[C@H](O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)56.0-66.0?C
FormPowder
Assay (GC)≥96.0%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only