Pyrimidine-5-boronic acid hemihydrate, 97%
Pyrimidine-5-boronic acid hemihydrate, 97%
Pyrimidine-5-boronic acid hemihydrate, 97%
Thermo Scientific Chemicals

Pyrimidine-5-boronic acid hemihydrate, 97%

CAS: 696602-91-2 | C8H12B2N4O5 | 265.827 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H29212.03
also known as H29212-03
Price (EUR)
87,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
87,20
Each
Chemical Identifiers
CAS696602-91-2
IUPAC Namebis((pyrimidin-5-yl)boronic acid) hydrate
Molecular FormulaC8H12B2N4O5
InChI KeyYXMKGFBYXBTOJS-UHFFFAOYSA-N
SMILESO.OB(O)C1=CN=CN=C1.OB(O)C1=CN=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
Proton NMRConforms to structure
FormPowder
Assay (Aqueous acid-base Titration)≥96.0%
It is employed as a intermediates for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only