3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 97%
3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 97%
3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 97%

CAS: 883738-38-3 | C18H27BN2O3 | 330.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H29224.03
also known as H29224-03
Price (EUR)
72,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
72,70
Each
Chemical Identifiers
CAS883738-38-3
IUPAC Name1-methyl-4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine
Molecular FormulaC18H27BN2O3
InChI KeyZYDMNNQZEUSDGG-UHFFFAOYSA-N
SMILESCN1CCN(CC1)C(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)86.0-92.0?C
FormPowder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only