2-Cyclohexylethylboronic acid pinacol ester, 96%
2-Cyclohexylethylboronic acid pinacol ester, 96%
2-Cyclohexylethylboronic acid pinacol ester, 96%
Thermo Scientific Chemicals

2-Cyclohexylethylboronic acid pinacol ester, 96%

CAS: 167692-95-7 | C14H27BO2 | 238.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31156.06
also known as H31156-06
Price (EUR)
173,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
173,00
Each
Chemical Identifiers
CAS167692-95-7
IUPAC Name2-(2-cyclohexylethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC14H27BO2
InChI KeyLDKDGYZAIIKCGK-UHFFFAOYSA-N
SMILESCC1(C)OB(CCC2CCCCC2)OC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4525-1.4585 @ 20?C
FormLiquid
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only