3-(1,4-Cyclohexadien-1-yl)-1-propanol, 97%
3-(1,4-Cyclohexadien-1-yl)-1-propanol, 97%
3-(1,4-Cyclohexadien-1-yl)-1-propanol, 97%
Thermo Scientific Chemicals

3-(1,4-Cyclohexadien-1-yl)-1-propanol, 97%

CAS: 87151-66-4 | C9H14O | 138.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31465.03
also known as H31465-03
Price (EUR)
76,90
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
76,90
Each
Chemical Identifiers
CAS87151-66-4
IUPAC Name3-(cyclohexa-1,4-dien-1-yl)propan-1-ol
Molecular FormulaC9H14O
InChI KeyDJJNAQALDCPMOU-UHFFFAOYSA-N
SMILESOCCCC1=CCC=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Impurity content3-Phenylpropanol content: <3.0% by NMR
FormLiquid
Assay (GC)>96.0%
Refractive Index1.5040-4.5090 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only