2,3-Dimethylthiophene, 97%
2,3-Dimethylthiophene, 97%
2,3-Dimethylthiophene, 97%
Thermo Scientific Chemicals

2,3-Dimethylthiophene, 97%

CAS: 632-16-6 | C6H8S | 112.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H31641.06
also known as H31641-06
Price (EUR)
324,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
324,00
Each
Chemical Identifiers
CAS632-16-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5160-1.5210 @ 20°C
It has also been found that 2,3-dimethylthiophene can be formylated to yield a dimethylthiophene aldehyde, which as expected proved to have the aldehyde group in the free a-position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has also been found that 2,3-dimethylthiophene can be formylated to yield a dimethylthiophene aldehyde, which as expected proved to have the aldehyde group in the free a-position.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

General References:

  1. S Gronowitz,; V Pavulans. The Signs of the Methyl-Methyl Spin Couplings in 2, 3-Dimethylthiophene, 2, 3-Dimethylfuran. Acta Chemica. 1966, 20 2515-2526.
  2. William J. King,; F. F. Nord. Studies In the Thiophene Series. V. Wolf-Kishner Reductions. J. Org. Chem. 1949, 14 (4), 638-642.