Iodine monochloride, 1M soln. in dichloromethane
Iodine monochloride, 1M soln. in dichloromethane
Iodine monochloride, 1M soln. in dichloromethane
Thermo Scientific Chemicals

Iodine monochloride, 1M soln. in dichloromethane

CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number H31938.AE
also known as H31938-AE
Price (EUR)
64,10
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
64,10
Each
Chemical Identifiers
CAS7790-99-0
IUPAC Nameiodochlorane
Molecular FormulaClI
InChI KeyQZRGKCOWNLSUDK-UHFFFAOYSA-N
SMILESClI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown
FormLiquid
Assay (Iodometric Titration)0.95-1.04M
Iodine monochloride is a useful reagent in organic synthesis and source of I+. It is used as a source of electrophilic iodine in the synthesis of certain aromatic iodides. It also cleaves C-Si bonds. Iodine monochloride will also add to the double bond in alkenes to give chloro-iodo alkanes. It can be dissolved in acetic acid to make Wijs solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only