(1S,2R)-(-)-cis-1-Amino-2-indanol, 97%
(1S,2R)-(-)-cis-1-Amino-2-indanol, 97%
(1S,2R)-(-)-cis-1-Amino-2-indanol, 97%
Thermo Scientific Chemicals

(1S,2R)-(-)-cis-1-Amino-2-indanol, 97%

CAS: 126456-43-7 | C9H12NO | 150.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H32066.03
also known as H32066-03
Price (EUR)
50,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
50,60
Each
Chemical Identifiers
CAS126456-43-7
IUPAC Name(1S,2R)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
Molecular FormulaC9H12NO
InChI KeyLOPKSXMQWBYUOI-BDAKNGLRSA-O
SMILES[NH3+][C@@H]1[C@H](O)CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-62.0 ± 2.0° (C=0.5 in chloroform)
Appearance (Color)White to cream
Melting Point (clear melt)113.0-123.0°C
FormPowder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only