2,2-Difluorocyclopropanemethanol, 97%
2,2-Difluorocyclopropanemethanol, 97%
2,2-Difluorocyclopropanemethanol, 97%
Thermo Scientific Chemicals

2,2-Difluorocyclopropanemethanol, 97%

CAS: 509072-57-5 | C4H6F2O | 108.088 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H32083.03
also known as H32083-03
Price (EUR)
151,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
151,00
Each
Chemical Identifiers
CAS509072-57-5
IUPAC Name(2,2-difluorocyclopropyl)methanol
Molecular FormulaC4H6F2O
InChI KeyXOLSMTBBIZDHSG-UHFFFAOYNA-N
SMILESOCC1CC1(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.3800-1.3855 @ 20°C
2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
RUO – Research Use Only