N-Boc-DL-proline, 98%
N-Boc-DL-proline, 98%
N-Boc-DL-proline, 98%
N-Boc-DL-proline, 98%
Thermo Scientific Chemicals

N-Boc-DL-proline, 98%

CAS: 59433-50-0 | C10H17NO4 | 215.249 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32347.06
also known as H32347-06
Price (EUR)
193,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
193,00
Each
Add to cart
Chemical Identifiers
CAS59433-50-0
IUPAC Name1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid
Molecular FormulaC10H17NO4
InChI KeyZQEBQGAAWMOMAI-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Proton NMRConforms to structure
Appearance (Color)White
Assay (Aqueous acid-base Titration)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only