4-Fluoro-3-nitrobenzonitrile, 97%
4-Fluoro-3-nitrobenzonitrile, 97%
4-Fluoro-3-nitrobenzonitrile, 97%
Thermo Scientific Chemicals

4-Fluoro-3-nitrobenzonitrile, 97%

CAS: 1009-35-4 | C7H3FN2O2 | 166.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32391.03
also known as H32391-03
Price (EUR)
31,90
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
31,90
Each
Chemical Identifiers
CAS1009-35-4
IUPAC Name4-fluoro-3-nitrobenzonitrile
Molecular FormulaC7H3FN2O2
InChI KeyLKOWKPGBAZVHOF-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC=C1F)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Pale yellow to yellow
Melting Point (clear melt)87.0-97.0?C
FormCrystals or powder or crystalline powder
4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only
T Furuya.; T Ritter. Carbon- fluorine reductive elimination from a high-valent palladium fluoride. Journal of the American Chemical Society.2008130 (31), 10060-10061.