Ethyl oxazole-5-carboxylate, 98%
Ethyl oxazole-5-carboxylate, 98%
Ethyl oxazole-5-carboxylate, 98%
Thermo Scientific Chemicals

Ethyl oxazole-5-carboxylate, 98%

CAS: 118994-89-1 | C6H7NO3 | 141.126 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32924.06
also known as H32924-06
Price (EUR)
76,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
76,60
Each
Chemical Identifiers
CAS118994-89-1
IUPAC Nameethyl 1,3-oxazole-5-carboxylate
Molecular FormulaC6H7NO3
InChI KeyKRMORCCAHXFIHF-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CN=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4660-1.4700 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.

Solubility
Soluble in water (35 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Min Wang.; Dengke Li.; Wei Zhou.; Lei Wang. A highly efficient palladium-catalyzed desulfitative arylation of azoles with sodium arylsulfinates. Tetrahedron 2012, 68 (7), 1926-1930.
  2. Chengliang Li.; Pinhua Li.; Jin Yang.; Lei Wang. Palladium-catalyzed deamidative arylation of azoles with arylamides through a tandem decarbonylation-C-H functionalization Chem. Commun. 2012, 48 (35 ), 4214-4216.