4,4-Dimethyl-2-pyrrolidinone, 95%
4,4-Dimethyl-2-pyrrolidinone, 95%
4,4-Dimethyl-2-pyrrolidinone, 95%
Thermo Scientific Chemicals

4,4-Dimethyl-2-pyrrolidinone, 95%

CAS: 66899-02-3 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33209.MD
also known as H33209-MD
Price (EUR)
119,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
119,00
Each
Chemical Identifiers
CAS66899-02-3
IUPAC Name4,4-dimethylpyrrolidin-2-one
Molecular FormulaC6H11NO
InChI KeyHGVPKAGCVCGRDQ-UHFFFAOYSA-N
SMILESCC1(C)CNC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale yellow
Melting Point (clear melt)41.0-53.0?C
Assay (GC)≥94.0%
FormCrystals or powder or crystalline powder or fused solid
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. C. Elisabet Tranberg.; Andrea Zickgraf.; Brian N. Giunta.; Henning Luetjens.; Heidi Figler.; Lauren J. Murphree.; Ruediger Falke.; Holger Fleischer.; Joel Linden.; Peter J. Scammells.; Ray. A. Olsson. 2-Amino-3-aroyl-4,5-alkylthiophenes:  Agonist Allosteric Enhancers at Human A1 Adenosine Receptors.J. Med. Chem. 2002, 45 (2),382-389 .
  2. Robert H. Dodd.; Catherine Ouannes.; Malka Robert-Gero.; Pierre Potier. Hybrid molecules: growth inhibition of Leishmania donovani promastigotes by thiosemicarbazones of 3-carboxy-.beta.-carbolines.J. Med. Chem. 1989, 32 (6),1272-1276 .