11-Azidoundecyltrimethoxysilane, 94%
11-Azidoundecyltrimethoxysilane, 94%
11-Azidoundecyltrimethoxysilane, 94%
Thermo Scientific Chemicals

11-Azidoundecyltrimethoxysilane, 94%

CAS: 334521-23-2 | C14H31N3O3Si | 317.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34298.MD
also known as H34298-MD
Price (EUR)
105,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
105,00
Each
Chemical Identifiers
CAS334521-23-2
IUPAC Name(11-azidoundecyl)trimethoxysilane
Molecular FormulaC14H31N3O3Si
InChI KeyACSJXAKDVNDMME-UHFFFAOYSA-N
SMILESCO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥92.0%
Refractive Index1.4430-1.4500 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only