1,3,5,7-Tetramethyl-6-phenyl-2,4,8-trioxa-6-phosphaadamantane
1,3,5,7-Tetramethyl-6-phenyl-2,4,8-trioxa-6-phosphaadamantane
1,3,5,7-Tetramethyl-6-phenyl-2,4,8-trioxa-6-phosphaadamantane
Thermo Scientific™

1,3,5,7-Tetramethyl-6-phenyl-2,4,8-trioxa-6-phosphaadamantane

CAS: 97739-46-3 | C16H21O3P | 292.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2 g
Catalog number H36051.04
also known as H36051-04
Price (EUR)
206,00
Each
Quantity:
2 g
Request bulk or custom format
Price (EUR)
206,00
Each
Chemical Identifiers
CAS97739-46-3
IUPAC Name1,3,5,7-tetramethyl-8-phenyl-2,4,6-trioxa-8-phosphatricyclo[3.3.1.1³,⁷]decane
Molecular FormulaC16H21O3P
InChI KeyAVVSJWUWBATQBX-UHFFFAOYSA-N
SMILESCC12CC3(C)OC(C)(CC(C)(O1)P3C1=CC=CC=C1)O2
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline solid
Assay by NMR≥98.0% (P-31 NMR)
Proton NMRConforms
Appearance (Color)White
Phosphorus-31 NMRConforms