Sand, washed
Sand, washed
Sand, washed
Thermo Scientific Chemicals

Sand, washed

CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
5000 g
Catalog number H37933.0I
also known as H37933-0I
Price (EUR)
149,00
Each
Quantity:
5000 g
Request bulk or custom format
Price (EUR)
149,00
Each
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormGranular powder or solid
Impurity contentFe2O3 ≤0.30%
Loss on ignition≤0.5%
Particle Size≤4.0% is ≥500um
Appearance (Color)Pale cream to pale brown
View more
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Joseph Wang; Robin Krause; Kirstin Block; Mustafa Musameh; Ashok Mulchandani; Michael J.Schoning. Flow injection amperometric detection of OP nerve agents based on an organophosphorus-hydrolase biosensor detector. Biosensors and Bioelectronics. 2003, 18, (2-3),255-260
  2. Angus I.Kingon; Jon-Paul Maria; S.K.Streiffer. Alternative dielectrics to silicon dioxide for memory and logic devices. Nature. 2000, 406, 1032-1038