5-Bromo-7-azaindole, 96%, Thermo Scientific Chemicals
5-Bromo-7-azaindole, 96%, Thermo Scientific Chemicals
5-Bromo-7-azaindole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-7-azaindole, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H50040.03
also known as H50040-03
1 g
Catalog number H50040.03
also known as H50040-03
Price (EUR)
24,50
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
24,50
Each
Chemical Identifiers
CAS183208-35-7
IUPAC Name5-bromo-1H-pyrrolo[2,3-b]pyridine
Molecular FormulaC7H5BrN2
InChI KeyLPTVWZSQAIDCEB-UHFFFAOYSA-N
SMILESBrC1=CN=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only