1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine, 98%
1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine, 98%
1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine, 98%
Thermo Scientific Chemicals

1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine, 98%

CAS: 231953-40-5 | C11H13ClF3N3 | 279.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50263.06
also known as H50263-06
Price (EUR)
344,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
344,00
Each
Specifications
CAS231953-40-5
Chemical Name or Material1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Formula Weight279.69
Health Hazard 1H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only