1-(4-Pyridyl)ethylamine, 97%
1-(4-Pyridyl)ethylamine, 97%
1-(4-Pyridyl)ethylamine, 97%
1-(4-Pyridyl)ethylamine, 97%
Thermo Scientific Chemicals

1-(4-Pyridyl)ethylamine, 97%

CAS: 50392-78-4 | C7H11N2 | 123.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50303.03
also known as H50303-03
Price (EUR)
85,85
Online Exclusive
101,00
Save 15,15 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
85,85
Online Exclusive
101,00
Save 15,15 (15%)
Each
Add to cart
Chemical Identifiers
CAS50392-78-4
IUPAC Name(1S)-1-(pyridin-4-yl)ethan-1-aminium
Molecular FormulaC7H11N2
InChI KeyHIZMJYQEHFJWQY-LURJTMIESA-O
SMILESC[C@H]([NH3+])C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to orange
FormViscous liquid or liquid
Assay (unspecified)≥96.0%
1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Arthur J. Ashe. Trimethylsilyl migrations in the 5-trimethylsilylbicyclo[2.1.0]pentanes. J. Am. Chem. Soc. 1973, 95, (3), 818-821.
  2. Hiroshi. Ichikawa.; Michihiko. Ogata. Molecular orbital approach to the interpretation of organic mass spectra. III. Determination of intermediate ion structure in the mass spectrometry of heteroaromatic compounds. J. Am. Chem. Soc. 1973, 95, (3), 806-811.