4,6-Dichloro-2-methylpyrimidine, 97%
4,6-Dichloro-2-methylpyrimidine, 97%
4,6-Dichloro-2-methylpyrimidine, 97%
Thermo Scientific Chemicals

4,6-Dichloro-2-methylpyrimidine, 97%

CAS: 1780-26-3 | C5H4Cl2N2 | 163.001 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50493.03
also known as H50493-03
Price (EUR)
53,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
53,60
Each
Chemical Identifiers
CAS1780-26-3
IUPAC Name4,6-dichloro-2-methylpyrimidine
Molecular FormulaC5H4Cl2N2
InChI KeyFIMUTBLUWQGTIJ-UHFFFAOYSA-N
SMILESCC1=NC(Cl)=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormSolid or powder
Assay (GC)96.0% min
ClarityClear colorless to pale yellow solution in chloroform, no particulates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only