2-Oxocyclohexanepropionitrile, 99%
2-Oxocyclohexanepropionitrile, 99%
2-Oxocyclohexanepropionitrile, 99%
Thermo Scientific Chemicals

2-Oxocyclohexanepropionitrile, 99%

CAS: 4594-78-9 | C9H13NO | 151.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50722.03
also known as H50722-03
Price (EUR)
47,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
47,00
Each
Chemical Identifiers
CAS4594-78-9
IUPAC Name3-(2-oxocyclohexyl)propanenitrile
Molecular FormulaC9H13NO
InChI KeySPTVCXKSSRBTMN-UHFFFAOYNA-N
SMILESO=C1CCCCC1CCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
FormLiquid
Mass SpecConforms
Purity≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only