1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.
Solubility
Soluble in ethyl acetate, hexane and methanol.
Notes
Moisture sensitive. Incompatible with oxidizing agents, heat, light, ultraviolet radiation and free radical initiators.
RUO – Research Use Only
General References:
- Robert F.Cozzens; Robert B.Fox. Intramolecular triplet energy transfer in poly(1-vinylnaphthalene). J. Chem. Phys. 1969, 50, (4),1532
- C.E.Hoyle; T.L.Nemzek; A.Mar; J.E.Guillet. Time resolved fluorescence studies of Poly(N-vinylcarbazole), Poly(1-vinylnaphthalene), and 1,3-Bis(N-carbazolyl)propane. Macromolecules. 1978, 11, (2),429-431