5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester, Thermo Scientific Chemicals
5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester, Thermo Scientific Chemicals
5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H51117.06
also known as H51117-06
5 g
Catalog number H51117.06
also known as H51117-06
Price (EUR)
141,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
141,00
Each
Specifications
CAS1218790-44-3
Chemical Name or Material5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester
Formula Weight307.26
IUPAC Name1-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]piperidine
InChI KeyKDPVLCCGXRCQCV-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only