Specifications
CAS301652-23-3
Chemical Name or Material4-Methyl-2-(tri-n-butylstannyl)pyridine
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Formula Weight382.16
Health Hazard 1H301-H312-H315-H319-H372
View more