1-Methyl-2-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific Chemicals
1-Methyl-2-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific Chemicals
1-Methyl-2-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methyl-2-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H51523.03
also known as H51523-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H51523.03
also known as H51523-03
Price (EUR)
284,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
284,00
Each
Specifications
CAS105494-69-7
Chemical Name or Material1-Methyl-2-(tri-n-butylstannyl)imidazole
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Formula Weight371.15
Health Hazard 1H301-H312-H315-H319-H372
View more
1-Methyl-2-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials. It is an organostannic compound for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-2-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials. It is an organostannic compound for proteomics research.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
Gang Zhang; Ruifa Zong; Huan-Wei Tseng; Randolph P.Thummel. Ru(II) complexes of tetradentate ligands related to 2,9-di(pyrid-2‘-yl)-1,10-phenanthroline. Inorg. Chem. 2008, 47, (3),990-998