5-Methyl-2-(tri-n-butylstannyl)thiazole, 90+%
5-Methyl-2-(tri-n-butylstannyl)thiazole, 90+%
5-Methyl-2-(tri-n-butylstannyl)thiazole, 90+%
Thermo Scientific Chemicals

5-Methyl-2-(tri-n-butylstannyl)thiazole, 90+%

CAS: 848613-91-2 | C16H31NSSn | 388.201 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H51531.03
also known as H51531-03
Price (EUR)
314,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
314,00
Each
Specifications
CAS848613-91-2
Chemical Name or Material5-Methyl-2-(tri-n-butylstannyl)thiazole
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Formula Weight388.19
Health Hazard 1H301-H312-H315-H319-H372
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only