N-Boc-3-fluoro-L-phenylalanine, 95%
N-Boc-3-fluoro-L-phenylalanine, 95%
N-Boc-3-fluoro-L-phenylalanine, 95%
Thermo Scientific Chemicals

N-Boc-3-fluoro-L-phenylalanine, 95%

CAS: 114873-01-7 | C14H18FNO4 | 283.299 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51988.06
also known as H51988-06
Price (EUR)
336,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
336,00
Each
Specifications
Beilstein5347375
CAS114873-01-7
Chemical Name or MaterialN-Boc-3-fluoro-L-phenylalanine
Formula Weight283.3
IUPAC Name(2S)-3-(3-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only