4-(Boc-amino)-N-Fmoc-L-phenylalanine, 95%
4-(Boc-amino)-N-Fmoc-L-phenylalanine, 95%
4-(Boc-amino)-N-Fmoc-L-phenylalanine, 95%
Thermo Scientific Chemicals

4-(Boc-amino)-N-Fmoc-L-phenylalanine, 95%

CAS: 174132-31-1 | C29H30N2O6 | 502.567 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52031.03
also known as H52031-03
Price (EUR)
248,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
248,00
Each
Specifications
CAS174132-31-1
Chemical Name or Material4-(Boc-amino)-N-Fmoc-L-phenylalanine
Formula Weight502.57
IUPAC Name(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]propanoic acid
InChI KeyKVUAOWDVYMUKPE-VWLOTQADSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only