(S)-3-(Boc-amino)adipic acid 6-benzyl ester, 95%, Thermo Scientific Chemicals
(S)-3-(Boc-amino)adipic acid 6-benzyl ester, 95%, Thermo Scientific Chemicals
(S)-3-(Boc-amino)adipic acid 6-benzyl ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-3-(Boc-amino)adipic acid 6-benzyl ester, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H52187.03
also known as H52187-03
1 g
Catalog number H52187.03
also known as H52187-03
Price (EUR)
236,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
236,00
Each
Specifications
Beilstein8079168
CAS218943-30-7
Chemical Name or Material(S)-3-(Boc-amino)adipic acid 6-benzyl ester
Formula Weight351.4
IUPAC Name(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-6-oxo-6-phenylmethoxyhexanoic acid
View more
Starting materials for peptide synthesis, peptidomimetic and medicinal chemistry, protected amino acids, coupling reagents, linkers and resins, natural and unusual amino acids and building blocks used in peptide synthesis, peptidomimetic and medicinal chemistry. Reagents for PEGylation, Life science Research and biocatalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Starting materials for peptide synthesis, peptidomimetic and medicinal chemistry, protected amino acids, coupling reagents, linkers and resins, natural and unusual amino acids and building blocks used in peptide synthesis, peptidomimetic and medicinal chemistry. Reagents for PEGylation, Life science Research and biocatalysis.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only