(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, 98+%
(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, 98+%
(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, 98+%
Thermo Scientific Chemicals

(1R,2R)-Cyclohexane-1,2-dicarboxylic acid, 98+%

CAS: 46022-05-3 | C8H12O4 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52293.06
also known as H52293-06
Price (EUR)
355,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
355,00
Each
Specifications
CAS46022-05-3
Chemical Name or Material(1R,2R)-Cyclohexane-1,2-dicarboxylic acid
Formula Weight172.18
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Chiral dicarboxylic acid guests with a two C spacer (1R,2R)-cyclohexane-1,2-dicarboxylic acid and Boc-L-aspartic acid] gave CD-active species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chiral dicarboxylic acid guests with a two C spacer (1R,2R)-cyclohexane-1,2-dicarboxylic acid and Boc-L-aspartic acid] gave CD-active species.

Solubility
Soluble in acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Gianfranco Ercolani.The Origin of Cooperativity in Double-Wheel Receptors. Freezing of Internal Rotation or Ligand-Induced Torsional Strain?. Journal name. 2005, 7 (5), 803-805.
  2. Prof. Gianfranco Ercolani,; Dr. Luca Schiaffino. Allosteric, Chelate, and Interannular Cooperativity: A Mise au Point. TOC. 2008, 50 (8), 1762-1768.