Specifications
CAS1073338-92-7
Chemical Name or Material2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester
Formula Weight266.1
IUPAC Nameethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate
InChI KeyUMMKKZJPSPIDGN-UHFFFAOYSA-N
View more