2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester, 96%
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester, 96%
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester, 96%
Thermo Scientific Chemicals

2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester, 96%

CAS: 1073338-92-7 | C13H19BO5 | 266.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52312.MD
also known as H52312-MD
Price (EUR)
86,40
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
86,40
Each
Specifications
CAS1073338-92-7
Chemical Name or Material2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester
Formula Weight266.1
IUPAC Nameethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate
InChI KeyUMMKKZJPSPIDGN-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only