2-(Methylsulfonyl)benzeneboronic acid, 98%
2-(Methylsulfonyl)benzeneboronic acid, 98%
2-(Methylsulfonyl)benzeneboronic acid, 98%
2-(Methylsulfonyl)benzeneboronic acid, 98%
Thermo Scientific Chemicals

2-(Methylsulfonyl)benzeneboronic acid, 98%

CAS: 330804-03-0 | C7H9BO4S | 200.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52445.03
also known as H52445-03
Price (EUR)
127,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
127,00
Each
Add to cart
Chemical Identifiers
CAS330804-03-0
IUPAC Name(2-methanesulfonylphenyl)boronic acid
Molecular FormulaC7H9BO4S
InChI KeyMSQFFCRGQPVQRS-UHFFFAOYSA-N
SMILESCS(=O)(=O)C1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only