3-Chlorocarbonylbenzeneboronic anhydride, tech. 90%
3-Chlorocarbonylbenzeneboronic anhydride, tech. 90%
3-Chlorocarbonylbenzeneboronic anhydride, tech. 90%
Thermo Scientific Chemicals

3-Chlorocarbonylbenzeneboronic anhydride, tech. 90%

CAS: 332154-58-2 | C7H6BClO3 | 184.382 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52722.03
also known as H52722-03
Price (EUR)
80,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
80,60
Each
Specifications
CAS332154-58-2
Chemical Name or Material3-Chlorocarbonylbenzeneboronic anhydride
Formula Weight166.37
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only