(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%
(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%
(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%
Thermo Scientific Chemicals

(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%

CAS: 24716-93-6 | C7H13NO2 | 143.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52779.MD
also known as H52779-MD
Price (EUR)
119,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
119,00
Each
Specifications
CAS24716-93-6
Chemical Name or Material(1S,2S)-2-Aminocyclohexanecarboxylic acid
Formula Weight143.19
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Vilaivan, T. Pyrrolidinyl PNA with alpha/beta-Dipeptide Backbone: From Development to Applications. Acc. Chem. Res. 2015, 48 (6), 1645-1656.
  2. Ma, C. D.; Wang, C.; Acevedo-Vélez, C.; Gellman, S. H.; Abbott, N. L. Modulation of hydrophobic interactions by proximally immobilized ions. Nature 2015, 517 (7534), 347-350.