2-(Methoxymethyl)benzeneboronic acid, 97%
2-(Methoxymethyl)benzeneboronic acid, 97%
2-(Methoxymethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

2-(Methoxymethyl)benzeneboronic acid, 97%

CAS: 126617-98-9 | C8H11BO3 | 165.983 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52858.MD
also known as H52858-MD
Price (EUR)
38,60
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
38,60
Each
Chemical Identifiers
CAS126617-98-9
IUPAC Name[2-(methoxymethyl)phenyl]boronic acid
Molecular FormulaC8H11BO3
InChI KeyJHPPAFXQJZJGEP-UHFFFAOYSA-N
SMILESCOCC1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
2-(Methoxymethyl)benzeneboronic acid used as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methoxymethyl)benzeneboronic acid used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only