1,3-Diphenyl-2-propyn-1-ol, tech. 90%
1,3-Diphenyl-2-propyn-1-ol, tech. 90%
1,3-Diphenyl-2-propyn-1-ol, tech. 90%
Thermo Scientific Chemicals

1,3-Diphenyl-2-propyn-1-ol, tech. 90%

CAS: 1817-49-8 | C15H12O | 208.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H53374.03
also known as H53374-03
Price (EUR)
35,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
35,20
Each
Specifications
CAS1817-49-8
Chemical Name or Material1,3-Diphenyl-2-propyn-1-ol
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Density1.099
Flash Point>110°C (230°F)
View more
1,3-Diphenyl-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Dr. Yoshiaki Nishibayashi; Youichi Inada; Masato Yoshikawa; Prof. Dr. Masanobu Hidai; Prof. Dr. Sakae Uemura. Propargylation of aromatic compounds with propargylic alcohols catalyzed by a cationic diruthenium complex . Angewandte Chemie International Edition. 2003, 42, (13),1495-1498
  2. Roberto Sanz; Delia Miguel; Alberto Martinez; Julia M.Alvarez-Gutierrez; Felix Rodriguez. Bronsted acid catalyzed propargylation of 1,3-dicarbonyl derivatives. Synthesis of tetrasubstituted furans. Org. Lett. 2007, 9, (4),727-730