3,4-Dimethylhexane, 97%
3,4-Dimethylhexane, 97%
3,4-Dimethylhexane, 97%
Thermo Scientific Chemicals

3,4-Dimethylhexane, 97%

CAS: 583-48-2 | C8H18 | 114.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H53498.06
also known as H53498-06
Price (EUR)
90,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
90,60
Each
Specifications
Beilstein1718744
CAS583-48-2
Chemical Name or Material3,4-Dimethylhexane
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
Density0.72
View more
3,4-Dimethylhexane may be used to study the mechanism of addition of hydrogen to 1-butene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylhexane may be used to study the mechanism of addition of hydrogen to 1-butene.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Wing Tsang. Hydrocarbon decomposition. 6. Thermal decomposition of 3,4-dimethylhexane, 2,2,3-trimethylpentane, tert-butylcyclohexane, and related hydrocarbons. J. Phys. Chem. 1972, 76, (2),143-156
  2. M.Moller; W.Ritter; H.J.Cantow. Micro- and macroconformation of macromolecules. Polymer Bulletin. 1981, 4, (10),609-616