3-Bromobenzophenone, 97%
3-Bromobenzophenone, 97%
3-Bromobenzophenone, 97%
Thermo Scientific Chemicals

3-Bromobenzophenone, 97%

CAS: 1016-77-9 | C13H9BrO | 261.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H53505.03
also known as H53505-03
Price (EUR)
90,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
90,60
Each
Specifications
CAS1016-77-9
Chemical Name or Material3-Bromobenzophenone
EINECS Number213-808-5
Formula Weight261.12
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only