4-(Trifluoromethyl)styrene, 98%, stab.
4-(Trifluoromethyl)styrene, 98%, stab.
4-(Trifluoromethyl)styrene, 98%, stab.
Thermo Scientific Chemicals

4-(Trifluoromethyl)styrene, 98%, stab.

CAS: 402-50-6 | C9H7F3 | 172.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H53513.03
also known as H53513-03
Price (EUR)
136,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
136,00
Each
Specifications
Beilstein1863548
CAS402-50-6
Chemical Name or Material4-(Trifluoromethyl)styrene
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density1.165
View more
4-(Trifluoromethyl)styrene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)styrene is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Moisture & light sensitive. Incompatible with oxidizing agents, acids, bases, heat, ultravioloet radiation and free radical initiators.
RUO – Research Use Only

General References:

  1. Riccardo Giovannini; Paul Knochel. Ni(II)-catalyzed cross-coupling between polyfunctional arylzinc derivatives and primary alkyl iodides. J. Am. Chem. Soc. 1998, 120, (43),11186-11187
  2. Jette Kischel; Irina Jovel; Kristin Mertins; Alexander Zapf; Matthias Beller. A convenient FeCl3-catalyzed hydroarylation of styrenes. Org. Lett. 2006, 8, (1),19-22