n-Butyl 4-aminobenzoate, 99%
n-Butyl 4-aminobenzoate, 99%
n-Butyl 4-aminobenzoate, 99%
Thermo Scientific Chemicals

n-Butyl 4-aminobenzoate, 99%

CAS: 94-25-7 | C11H15NO2 | 193.246 g/mol
Have Questions?
Catalog NumberQuantity
H54690.22
also known as H54690-22
100 g
Catalog number H54690.22
also known as H54690-22
Price (EUR)
69,60
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
69,60
Each
Chemical Identifiers
CAS94-25-7
IUPAC Namebutyl 4-aminobenzoate
Molecular FormulaC11H15NO2
InChI KeyIUWVALYLNVXWKX-UHFFFAOYSA-N
SMILESCCCCOC(=O)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥98.5% (HPLC) (U.S. specification)
Appearance (Color)White
CommentSpecification differs for U.S. and non-U.S. material where indicated
Assay (GC)≥98.5% (non-U.S. Specification)
FormPowder
n-Butyl 4-aminobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butyl 4-aminobenzoate is used as pharmaceutical intermediate.

Solubility
Very slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hans R.Kricheldorf; Karsten Hauser. Polylactones. 55. A-B-A Triblock copolymers of various polypeptides. Syntheses involving 4-aminobenzoyl-terminated poly(ε-caprolactone) as B block. Biomacromolecules. 2001, 2,(4), 1110-1115. 
  2. Wolfgang Schrader; Jan T.Andersson. Fast and direct method for measuring 1-octanol-water partition coefficients exemplified for six local anesthetics. Journal of Pharmaceutical Sciences. 2001, 90,(12), 1948-1954.